When the number of FASTA indexes reaches the specified maximum, the application begins to automatically remove FASTA files from memory to not exceed the maximum value. The application first removes the oldest indexes (that is, the ones with the earliest access time). However, you can mark specific FASTA indexes so that they will not be removed from memory, even after the maximum is reached. To changed the number of maximum indexes, see Change the number and location of stored FASTA indexes.
Procedure
- Follow one of the procedures in the following table.
Task | Procedure |
|---|---|
Deactivate automatic FASTA index removal | Procedure
|
Activate automatic FASTA index removal | Procedure
|