The following table describes the options and columns in the FASTA Files view of the application.
Parameter | Description |
|---|---|
Opens the Open dialog box where you can select a FASTA database to import. | |
Opens the Download from the Annotation Server dialog box where you can download databases. | |
Updates the selected file from the Annotation Server if an updated file is available. This button is activated for downloaded FASTA files only if a new version of the file is available. | |
Deletes a FASTA database from the FASTA files view. | |
Cancels the addition or removal of a FASTA file. | |
Exports a FASTA file downloaded from the Annotation Server.
| |
Redisplays the view on the screen. | |
Checks to see if the data on the Protein Annotation server has been updated. | |
Name | Displays the name of the FASTA file. |
Protein Database | Specifies the name of the original source database from which the proteins were downloaded. |
Taxonomy ID | Displays the taxonomy identification number of the organism. |
Version | Displays the version of the FASTA file downloaded from the Protein Annotation server. |
File Size [kB] | Displays the current size of the FASTA file. |
# Sequences | Displays the number of sequences found in the FASTA file during processing. |
# Residues | Displays the number of amino acids found in the FASTA file during processing. |
Status | Displays the current status of the FASTA file. The statuses are as follows:
|
Last Modified | Displays the date when the FASTA file was last modified or created. |
Update Available | If selected, indicates that an updated database is available. |