You can list all the FASTA files that you have both downloaded from other sources onto your hard drive and then registered.
Procedure
  1. Do one of the following:
  2. From the application menu bar, select Administration > Maintain FASTA Files.
  3. Select Maintain FASTA Files .
  4. The FASTA Files view opens, which lists all the FASTA files that you have downloaded from other sources and registered. It displays the processed FASTA file properties, such as the file name, file size, and the number of proteins stored.
  5. The Proteome Discoverer application analyzes each protein entry to determine if the FASTA file meets the application requirements for use in a spectra search. It processes the FASTA file and makes it available for use.
FASTA Files view