After you perform a search, you can see the workflow and the analysis that the application used to produce the search results.
Procedure
- In an open study, select the Analysis Results tab.
- Right-click the name of the results file and then select Show Details.
- The Analysis Sequence Details window opens. It displays the Workflow Editor and the Analysis window that contains the processing workflows and consensus workflows used to generate the selected results file.