There are two methods of performing a search:
- The default mode, which creates one results file for all input files.
- The batch mode, which processes each file in a set of files with the same processing workflow and the same consensus workflow.
- To use batch mode, you must have more than one input file and only one processing step. (Workflows appear as steps in the Processing Step area in the Analysis pane).
- See Perform a search in each file separately.
NOTE
A highlighted node in the workflow indicates that the node is new or changed since the last search, and that you must run the search to update results. (Usually, the application can run the search on only those parts of the workflow that require it.)
Procedure
- Create a new study. See Create a study.
- In the Analysis window, select Run.