On the Grouping & Quantification page, you can:
- Select the study variables that were used to group your samples and select the numerators and denominators of your ratios.
- Manually specify the quantification ratios to generate or semiautomatically create all possible quantification ratios against an ad-hoc selection of denominator values.
For information about how the application internally handles grouping, see Understanding protein grouping.