These instructions apply to creating processing workflows and consensus workflows.
Procedure
- On the Start page, open a study by doing one of the following:
- Select Open Study.
- Select File > Open Study.
- The Open Study dialog box opens.
- Locate the study.
- Select Open.
- The study opens as a separate page in the Proteome Discoverer application.
TIP
You can also open a study by selecting a recent study from the Recent Studies column on the Start page.
- If an analysis is not already open, select New Analysis.
- On the Study page, select the Workflows tab.
- Do one of the following:
- Select the title bar of the Consensus Step area of the Analysis pane.
- A blue border highlights the Consensus Step area in the Analysis pane.
- Select the title bar of the Processing Step area of the Analysis pane.
- A blue border highlights the Processing Step area in the Analysis pane.
- On the Workflow Editor page, do one of the following:
- Select Open Common to open a workflow file that you choose.
- Select Open to open the most recent file path that you opened.
- If you clicked the Processing Step bar, the Open Workflow dialog box displays .pdProcessingWF files in the Common Templates folder.
- If you clicked the Consensus Step bar, the Open Workflow dialog box displays .pdConsensusWF files in the Common Templates folder.
- Select the workflow file, and select Open.
- The workflow opens in the Workflow Tree pane. If you must update the template, see Correct workflow errors.
- Configure the search node.
- Make any other changes required.
- Save the workflow as follows:
- In the Workflow box, enter a name for the workflow.
- (Optional) In the Description box above the Workflow Tree pane, enter a brief description of the workflow.
- Do one of the following:
- Select Save to save the current workflow as a template file.
- Select Save Common to save the current workflow as a template to the common templates folder. - The Save Workflow dialog box opens.
- In the Save Workflow dialog box, do the following:
- Navigate to the folder that you want to save the file to.
- Enter a name for the file. - Select Save.
- The application saves processing workflows as .pdProcessingWF files and saves consensus workflows as .pdConsensusWF files.