The following table describes the parameters available in the Analysis window.
Parameter | Description |
|---|---|
As Batch | Determines whether the application performs an analysis in batch mode. For information, see “Perform searches” on page 263. |
Tells the application to perform an analysis. | |
Opens Windows Explorer so that you can save the analysis as a .pdAnalysis template file. | |
Consensus Step box | Displays the name of the consensus workflow, the name of the consensus workflow results file, and the processing workflow that generated the MSF file that was submitted to the consensus workflow as an input file. |
Edit | Opens the Workflow Tree window that contains the workflow. |
Indicates that the consensus workflow contains an error. | |
Workflow | Displays the name of the consensus workflow. |
Result File | Displays the name of the file containing the results of the consensus workflow. |
Child Steps | Displays the processing workflow. |
Add | Adds an empty Processing Step box. |
Processing Step box | Displays the name of the processing workflow, the name of the processing workflow results file, and the input files that were submitted to the processing workflow as input files. |
Edit | Opens the Workflow Tree window containing the processing workflow. |
| Indicates that the processing workflow contains an error. |
Reprocess | Opens a locked processing workflow so that you can make changes to it. |
Clone | Opens a child step window so that you can create a different processing workflow for the same file or a different file. |
Workflow | Displays the name of the processing workflow. |
Result File | Displays the name of the file containing the processing workflow results. |
Input Files | Lists the input raw data files that were submitted to the processing workflow. |