From a selected analysis result, you can create a new analysis template that uses the same sequence steps as the selected result. This procedure removes the input files from the analysis and clears the result file names if they were automatically generated from the input files.
Procedure
- Select the Analysis Results tab.
- Select the appropriate analysis result on the page.
- Select Reprocess and then select Use as Analysis Template.