You can perform only the processing step in a workflow, you can remove the consensus step and re-add it later. For example, you only need the processing step to export peak lists as an MGF file with the Spectrum Exporter node.
NOTE
This process is only used when exporting the processing step to an MGF file or an mzML file.
Task | Procedure |
|---|---|
Remove the consensus step from an analysis | Procedure
|
Add the consensus step to an analysis | Procedure
|