In an analysis, a processing step is a step where the application runs a processing workflow on the input files. An analysis can contain more than one processing step. You can use additional processing steps to process one set of input files differently from another set of input files. For example, you can run a Percolator validation separately on a set of files, such as the replicates of the control group and the replicates of the treatment group.
Task | Procedure |
|---|---|
Add multiple processing steps | Procedure
|
Duplicate an existing processing step and its workflow | Procedure
|
NOTE
If you are increasing the number of processing steps, verify that you can run more processes in parallel. See Specify the number of workflows to execute in parallel.