When you create an analysis, two new tabs appear on the Study: Study name page: the Workflows tab and the Grouping & Quantification tab.
An Analysis window contains the following items:
- A Consensus Step box, which represents the consensus workflow step of the data processing.
- A Processing Step box, which represents the processing workflow step of the data processing.
- A Child Steps bar, which contains an Add button that you can use to add another Processing Step box.
TIP
Multiple processing steps are useful when you want to improve parallelization for workflows with single-threaded nodes or when different data files need to be processed differently.