An analysis in the Proteome Discoverer application is the actual data processing in the context of a study. An analysis consists of the different data processing and reporting steps to perform their associated workflows, their parameter settings, their input files, and other information related to how your data is being processed. A study can contain multiple analyses.
NOTE
A .pdAnalysis file contains one or more processing workflows and one consensus workflow.