You can use the Proteome Discoverer application to process Thermo Fisher Scientific raw data collected from the following mass spectrometers: Hybrid mass spectrometers and Tribrid mass spectrometers. Thermo Fisher Scientific also supports open formats such as MGF, mzData, mzXML, and mzML with limited functionality.