Label-free quantification is the quantification of MS peptide signals in LC/MS/MS data without an isotopic labeling reagent. Label-free quantification uses the Minora algorithm to perform untargeted feature detection, and it does not require an identification to quantify a peptide or protein. In theory, label-free quantification quantifies the most peptides and provides the widest dynamic range for untargeted workflows. It requires only a single PSM to quantify across all samples when you use retention-time alignment and feature mapping.
Label-free quantification proceeds as follows:
- First, the algorithm detects LC/MS peaks in the individual raw data files and maps them to identified PSMs, when available.
- Next, it creates new features from peaks that were not previously associated with any other feature. It takes all LC/MS peaks within a small retention-time range and checks for peaks that build isotope patterns.