The Proteome Discoverer application uses a study management system to organize your experiment data and to automate processing. The study management system comprises studies and analyses. An analysis contains the processing and consensus workflows that you assemble in the Workflow Editor to process your data.
A study contains the following:
  • Input files
  • Information about the samples contained in the files
  • Information about the treatment of the samples
  • Workflows used to process the input files
  • Results of the processing