The following table contains known defects in the software, categorized by software section, with a brief abstract and information related to each defect’s severity and risk. The Item ID is the internal number assigned to each issue. Product management assesses risk, which can differ significantly from the reported severity.
Software section | Severity | Abstract | Risk | Item ID |
|---|---|---|---|---|
XlinkX | High | Crosslinking: XlinkX open search still errors. | High | 403291 |
XlinkX | High | Crosslinking: XL cannot perform the analysis with the DSSO raw file acquired by the Astral instrument. | High | 418219 |
PD powered by Ardia | High | PD server crashes after a network loss. | Low | 363879 |
CHIMERYS | High | A queued job was started in the CHIMERYS server but its analysis fails when the PC automatically restarts for Windows updates. | Low | 387738 |
TMT | High | TMT processing workflow fails for specific raw files. | Low | 406083 |
XlinkX | Medium | XL Sequence not the same between the Crosslinks table and its associated CSMs table. | Low | 268036 |
Spectrum Grouper | Medium | There seems to be a performance issue in Spectrum Grouper node. | Low | 336580 |
Heat map | Medium | Out-of-memory in Heatmap. | Low | 400330 |
LFQ | Medium | Peptide abundances are changed after PD3.1.0.475. | Low | 402644 |
PD powered by Ardia | Medium | Importing a study from .csv does not work when remote files are included. | Low | 403802 |
CHIMERYS | Medium | Large Datasets: CHIMERYS analysis fails with local fractions if the input and processing mode is set to One job for All files. | Low | 405844 |
Precursor Detector | Medium | Protein Score is calculated wrong when using precursor detector node. | Low | 408138 |
Mascot | Medium | Context Help should work in the Mascot Communication tester the same as it works in the CHIMERYS Communication tester. | Low | 411174 |
Daemon | Medium | Cannot run CHIMERYS Ardia workflows using the Daemon utility because the identity token is missing. | Low | 412435 |
PD powered by Ardia | Medium | Inconsistency between processing statistics and job queue. | Low | 424170 |
Fragment Match Spectrum | Medium | The scale of x-axis of the spectrum matching was much bigger than the fragments ion coverage area under both Reference Spectrum checked and unchecked. | Low | 266072 |
XlinkX | Low | Noncleavable_fast analysis templates do not work in PD 3.1. | Low | 427509 |
Thirdparty installer | Low | The release notes link is not working in the thirdparty installer. | Low | 427551 |