The following table lists defects that were resolved between Proteome Discoverer 3.0 and Proteome Discoverer 3.1. The table excludes Help issues and any cosmetic fixes. In some cases the Abstract has been amended or extended from the original to better describe the reported issue. Both an engineering fix and follow-up testing (verified by our product evaluation department) have resolved each of these issues.
Item ID
Software section
Abstract
249161
CHIMERYS
CHIMERYS search failed because the percolator output cannot be obtained from CHIMERYS.
309299
Percolator
Percolator node failed in a PWF because the root element is missing.
336547
Minora
Minora node crashes in some cases (when a RAW file is registered internally with a different file size).
400386
Minora
Minora crashes when processing multiple RAW files in parallel.
423780
Data distributions
Found in distributions does not show the correct label for low confident PSMs.
309504
Sequest HT
Sequest HT crashes if an amino acid is specified twice in the cleavage site description of an enzyme.
324577
Statistics
User can create inverse ratios of each other.
281300
Modification editor
Not all the Neutral Losses and Diagnostic Ions should be deleted after importing the Neutral Losses and Diagnostic Ions from a .xml file.
357141
Comet
There is no annotation on the Fragment Spectrum plot.
359125
CHIMERYS
I/L problem with CHIMERYS.
361249
CHIMERYS
Enhancement request: Give users a clear warning that tells them to re-login to the CHIMERYS server after being logged into the server for more than 60 days. The user can do this either at the parameter validation stage or in Job queue.
371895
Fragment spectrum
The default color of 1st spectrum in the mirror plot is yellow.
308331
Spectrum Files RC
Enhancement request: Show all of the default setting in Static Modifications for the Spectrum files RC node without needing to open the Show Advanced Parameters.
309247
Log files
Operating System ID in Logfiles points to an old Win8 version.
341275
FASTA databases
Enhancement request: Let users update multiple databases with one click.
348792
Summary information
A max Mass of a Raw file is not correct in the Input files table of a pdResult.