If you have an Ardia server in your laboratory, a license key will be sent to you that activates the Ardia features in Proteome Discoverer. Once the license key is entered, you may register an Ardia server to Proteome Discoverer. Once registered to an Ardia, you may load remote injections into studies and publish results from studies to the Ardia server. You can also register a single Proteome Discoverer installation for automated analysis. For more information, please consult the User Guide.
If you own an Ardia Advanced Tower PC, we will also send you a license key to activate the CHIMERYS on Ardia Server node. This node allows submission of CHIMERYS searches to the local PC rather than via the cloud service.