Proteome Discoverer 3.1 can connect to the Ardia™ Platform to access raw files and to publish Proteome Discoverer results. When connected to the Ardia Advanced Tower Server, Proteome Discoverer has the following additional features:
  • CHIMERYS on Ardia Server: The Ardia Advanced Tower Server hosts a local version of the CHIMERYS algorithm, bypassing the need to send data to the service hosted in Amazon Web Services.
  • Automation: If Xcalibur™ 4.7 data system is registered to the same Ardia server as a PC with Proteome Discoverer 3.1, the Proteome Discoverer 3.1 PC can be configured for automated analysis. Proteome Discoverer 3.1 lets you choose specific analyses to publish to Xcalibur through the Ardia server. These analyses can be selected in a sequence along with a quantification method and a target folder for the results of processing. After Xcalibur finishes acquisition of each sample, the file will be automatically uploaded to the Ardia server and subsequently sent to the Proteome Discoverer data analysis PC for analysis. Once the analysis is complete, the result file will be uploaded to the Ardia server to the folder selected in the Xcalibur sequence.
For more information on the other features of the Ardia Platform, visit https://www.thermofisher.com/us/en/home/digital-solutions/ardia-platform.html.