View Sample Abundances - Proteome Discoverer Software - View Sample Abundances - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Open the Report Item Distribution view by either selecting View > Distribution Charts or Report Item Distribution
. - In the Report Item Distribution view, select the Sample Abundances tab.
- For Data Source, select Proteins: Abundances (Normalized).