Use the display filter to view E.coli proteins - Proteome Discoverer Software - Use the display filter to view E.coli proteins - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
Product Tree
Desktop Software > Proteome Discoverer Software
Ardia Platform > Software > Proteome Discoverer Software
Document Type
User Guide
Software version
3.1
Select the Proteins page in the result file. Select the Display Filter . The Display Filter pane opens. In the Display Filter pane, do the following: Select Add Property , and then select Description . Keep the red box selection Contains . Enter Escherichia in the text box. Select Apply at the top of the Display Filters pane. All of the Description entries contain Escherichia, which indicates that these are E. coli proteins. Scroll the Proteins table to the right until the Abundance Ratios column is visible.