Procedure
- Sort the Proteins table by selecting the Sum PEP Score column.
NOTE
The human proteins are still filtered out in this list from the steps in Use the display filter to view E. coli proteins.
- Select the protein DNA-directed RNA polymerase subunit beta in the first row.
- Select Show Associated Tables at the bottom of the window.
- Select the Peptide Groups table if it is not already selected in the Associated Table.
- Select View > Chromatogram Traces View to view the chromatograms for the selected peptide.
NOTE
By default, the chromatograms are color-coded by their study factor. You can change the study factor with the Group By checkboxes on the left side of the Chromatogram Traces view. When you select the study factor, the three replicates of each sample appear in the same color. As you can see by the plot, the chromatograms for the 196 ng sample are more abundant than the 92 ng by ~2X. This shows that the quantification results match the expectation for an E. coli peptide.
- Select the Annotated Sequence column.
- Select the peptide LIPAGTGYAYHQDR.
- The chromatogram traces view updates with the chromatograms for that specific peptide.
- The chromatographic peaks for this peptide are well aligned.
- The vertical lines on the plot show when that peptide was triggered for MS/MS during acquisition for that file.
- Also, triangles are shown at the apex of each chromatogram. The Precursor Ions Quantifier node allows changing the setting for peptide quantification between the apex intensity indicated by that triangle or the peak area indicated by the area shaded under the chromatogram.
- Select the field chooser and add the LCMS/MS Features table to the results.