Use the display filter to view protein quan occupancy - Proteome Discoverer Software - Use the display filter to view protein quan occupancy - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
Product Tree
Ardia Platform > Software > Proteome Discoverer Software
Desktop Software > Proteome Discoverer Software
Document Type
User Guide
Software version
3.1
Select the Proteins table in the result file. Select the Display Filter . The Display Filter pane opens. In the Display Filter pane, do the following: Select Add property and then select Abundances . In the pink box to the right of the property, select has any value . In the second pink box, select in every sample . The filtered results show 8892 out of 9720 protein groups were quantified in every sample or 90% of proteins.