You already selected a processing workflow in the New Study and Analysis dialog box. In this section, you configure some settings in the workflow nodes.
Procedure
- Select the Workflows tab.
- In the Analysis pane, select Edit in the header of the Processing Step.
- In the Workflow Tree, select the Spectrum Files RC node.
- For the Protein Database parameter, select the yeast FASTA file, Saccharomyces cerevisiae.
- This is the file you added in Add the FASTA file to Proteome Discoverer.
- Select the Sequest HT node in the Workflow Tree.
- For the Protein Database parameter, select the yeast FASTA file, Saccharomyces cerevisiae.