The study holds the study factors, raw data files, quantification methods, and links to all search results for analyses performed on those data files within the study.
Procedure
- Select the Start Page tab in the Proteome Discoverer application.
- In the Start window, select New Study/Analysis.
- The New Study and Analysis dialog box opens.
- In the New Study and Analysis dialog box, do the following:
- In the Study Name text field, enter a name (for example, Triple Knockout Two Replicates).
- (Optional) For Study Root Directory, change the folder for storing the study.
- Keep the study on a local drive.
- Select the Processing workflow:
- ProcessingWF_Tribrid\ PWF_Tribrid_TMT_Quan_SPS_MS3_SequestHT_Percolator.pdProcessingWF
- Select the Consensus workflow:
- ConsensusWF\CWF_Comprehensive_Enhanced Annotation_Reporter_Quan.pdConsensusWF
- Select OK.
- In the Quantification Methods section of the Study Definition page, select Add, and select TMT 11plex SG253828.
- This is the method you created in Configure the quantification method.
- Define the study factors, and associate them with the quantification channels.
- In the Study Factors section of the Study Definition pane, select Add > Categorical Factor.
- For the title of the factor, enter Yeast Strain, and select Apply.
- In the New Factor dialog box, select Edit.
- Add the following items to the new study factor: Met6, His4, Ura2, and Parental.
- Select Apply.
- Select the Input Files tab, and then the select Add Files.
- The Add Files dialog box opens.
- Select both of the TMT 11plex data files (TKOTT11_1ms3_1 and TKOTT11_1ms3_2), and then select Open.
- In the Quan Method column next to the file name, select the TMT 11plex SG253828 quan method for both data files.
- Press Shift and select both files, and then drag them from the Input Files tab to the Files for Analysis section of the Analysis.
- The files are now part of the analysis.
- Select the Samples tab.
- In the Yeast Strain column, make the following changes in the quantification for both data files:
- Change the 126, 127N, and 127C quan channels to Met6.
- Change the 128N, 128C, and 129N quan channels to His4.
- Change the 129C, 130N, and 130C quan channels to Ura2.
- Change the 131N and 131C columns to Parental.
TIP
You can enter these changes one row at a time using the dropdown list, or by selecting multiple rows, right-clicking, and in the dropdown list selecting Set Yeast Strain to.
- For the four parental channels, change the Sample Type column from Sample to Control.