Download the FASTA file from the UniProt website - Proteome Discoverer Software - Download the FASTA file from the UniProt website - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
- Product Tree
- Desktop Software > Proteome Discoverer Software
- Ardia Platform > Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Go to https://www.uniprot.org/.
- Select Proteomes.
- In the search field, enter Saccharomyces cerevisiae, and select Search.
- Select UP000002311.
- On the left side of the page, select Reviewed.
- Select Download.
- In the dialog box, do the following:
- Keep the Download All option.
- For Format, select Format FASTA (canonical & isoform).
- Select the Uncompressed option, and select Go.
- Save the file.
- After the file download is complete, on the Windows desktop, double-click the Proteome Discoverer application.
- On the Start Page, select Administration > Maintain FASTA files.
- In the FASTA Files view, select Add.
- Select the file that you downloaded from UniProt.
- The application indexes the file and adds it to the list of available databases.
- Continue with Configure the quantification method.