There are several methods of accessing the results.
Procedure
- Do one of the following:
- Select the Analysis Results tab in the study, and do one of the following:
- Double-click the row with the result ID 1.
- Select the row, and then select Open Result to open the result file in a separate tab. - Open the Job Queue by selecting Administration > Show Job Queue, and do one of the following:
- Double-click the row in the Job Queue for the Consensus workflow that just finished.
- Highlight the row, and then select Open Result to open the results in a separate tab. - In File Explorer, find the study folder, and then open the .pdResult file.