This familiarization tutorial provides step-by-step instructions for using the Proteome Discoverer application's precursor ion quantification capabilities.
This familiarization exercise uses SILAC-based quantitation. The sample used in this exercise was acquired using Thermo Scientific Orbitrap Elite™ Hybrid Ion Trap-Orbitrap Mass Spectrometer.
For general information about SILAC-based quantitation, go to https://www.analyteguru.com/.
|
|
---|---|
Quan method: | SILAC 3plex (Arg6 Lys4, Arg10 Lys8) |
Quan channels: | Light, medium, heavy |
Organism: | E. coli |
Instrument: | Orbitrap Elite |
Acquisition method: | Data dependent MS/MS, HCD |
RAW file: | Chris_Ecoli_1-2-4.raw Located in the application media in the folder: Example RAW Files. Copy to a local directory. |
Sample Description: | The experiment and labeling approach aimed at 1:2:4 ratios for lysine and 4:2:1 ratios for arginine. |
Disk space: | 1GB local drive space, SSD drive preferred |
Processing time: | Approximately 10 minutes |