Set up the ratio and complete the analysis - Proteome Discoverer Software - Set up the ratio and complete the analysis - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
Product Tree
Desktop Software > Proteome Discoverer Software
Ardia Platform > Software > Proteome Discoverer Software
Document Type
User Guide
Software version
3.1
In the Study, select the Grouping & Quantification tab. Under the Study Variables pane, select the E coli (ng) option. In the Bulk Ratio Generation pane, select E coli (ng): 96 and E coli (ng): 192 . Select Add Ratios . There are now two ratios (192/96) in the Generated Ratios pane. In the Analysis pane, select Run . The processing and consensus workflows appear in the Job Queue.