Download the FASTA file from UniProt website - Proteome Discoverer Software - Download the FASTA file from UniProt website - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
- Product Tree
- Desktop Software > Proteome Discoverer Software
- Ardia Platform > Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- Go to https://www.uniprot.org/.
- Select Proteomes.
- To download the Homo sapiens FASTA file:
- In the search field, enter homo sapiens, and select Search.
- Select UP000005640.
- On the left side of the page, select Reviewed.
- Select Download.
- In the dialog box, do the following for the Homo sapiens FASTA file:
- Keep the Download All option.
- For Format, select Format FASTA (canonical & isoform).
- Select the Uncompressed option, and select Go.
- Save the Homo sapiens FASTA file.
- To download the E. coli FASTA file:
- In the search field, enter escherichia coli, and select Search.
- Select UP000000625.
- On the left side of the page, select Reviewed.
- Select Download.
- In the dialog box, do the following for the E. coli FASTA file:
- Keep the Download All option.
- For Format, select Format FASTA (canonical & isoform).
- Select the Uncompressed option, and select Go.
- Save the E. coli FASTA file.
- After the E. coli file download is complete, open the Proteome Discoverer 3.1 application.
- On the Start Page, select Administration > Maintain FASTA files.
- In the FASTA Files view, select Add.
- Select the file that you downloaded from UniProt.
- The application indexes the file and adds it to the list of available databases.
- Continue with Create the study.