Procedure
  1. On the Windows desktop, double-click the Proteome Discoverer application.
  2. On the Start Page, select Administration > Maintain FASTA files.
  3. In the FASTA Files View, if you have not downloaded FASTA files to the application before, select Check For Updates.
  4. The Job Queue opens and runs a process that downloads the annotation database from Annotation Server. Wait for this step to complete before proceeding.
  5. To return to the FASTA Files view, select FASTA Files under Content Management.
  6. Select Download.
  7. In the Download from Annotation Server dialog box, do the following.
  8. For Taxonomy ID, enter 9606.
  9. Ensure that Include all subtaxonomies checkbox is clear.
  10. For Database, select the sp_incl_isoforms database at the top of the list.
  11. Select Import.
  12. The Run Queue runs a new process that downloads the human protein database. Depending on your network speed, this process might take a few minutes to complete.
  13. In the Download from Annotation Server dialog box, do the following.
  14. For Taxonomy ID, enter 83333.
  15. Ensure that Include all subtaxonomies checkbox is clear.
  16. For Database, select the sp_canonical database at the top of the list.
  17. Select Import.