Set up the ratios and complete the analysis - Proteome Discoverer Software - Set up the ratios and complete the analysis - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
Product Tree
Ardia Platform > Software > Proteome Discoverer Software
Desktop Software > Proteome Discoverer Software
Document Type
User Guide
Software version
3.1
In the Study, select the Grouping & Quantification tab. Under the Study Variables section, clear the File and Quan Channel options. Under the Study Variables section, select the E coli (ng) option. In the Bulk Ratio Generation pane, select E coli (ng): 10 , and then select Add Ratios . There are now three ratios (3/10, 7.5/10, 15/10) in the Generated Ratios pane. In the Analysis pane, select Run . The processing and consensus workflows appear in the Job Queue.