Set up the consensus workflow - Proteome Discoverer Software - Set up the consensus workflow - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
Product Tree
Ardia Platform > Software > Proteome Discoverer Software
Desktop Software > Proteome Discoverer Software
Document Type
User Guide
Software version
3.1
Open the consensus workflow by selecting Edit to the right of the Consensus Step within the Analysis section. Select the Precursor Ions Quantifier Node and do the following: For Normalization Mode, select Specific Protein Amount . For Proteins for Normalization, select the Homo sapiens FASTA database. Select the Protein Annotation node. For the Protein Database, select the Homo sapiens database and the E. coli FASTA database from the dropdown list.