Procedure
- Select the Workflows tab.
- Select Edit next to the Processing Step within the Analysis section.
- Select the Spectrum Files RC node in the Workflow Tree.
- For the Protein Database parameter, select the human FASTA file and the E. coli FASTA file.
- These are the files you added in Add the FASTA files to Proteome Discoverer.
- Select the Sequest HT node #4 in the Workflow Tree.
- For the Protein Database parameter, select the human FASTA file and the E. coli FASTA file.
- Select the Sequest HT node #8 in the Workflow Tree.
- For the Protein Database parameter, select the human FASTA file and the E. coli FASTA file.
- Select the Workflow Nodes tab on the lower left of the Workflow Editor.
- Under Feature Detection & Quantification, select and drag the Minora Feature Detector node to the side of the Spectrum Files RC node in the Workflow Tree.
- The node automatically connects to the Spectrum Files RC node.
NOTE
If using the CHIMERYS workflow, you will need to choose the Human and E coli databases, ensure that inferys_2_1_fragmentation is selected for the Prediction Model, Oxidation (M) is selected as a dynamic modification, and Carbamidomethyl (C) is selected as a static modification. The Minora Feature Detector is already in the workflow and thus does not need to be added.