This exercise uses the TMT 10plex quantification method.
Procedure
- Select Administration > Maintain Quantification Methods.
- In the Quantification Methods view toolbar, select Add to create a new quantification method.
- In the Create New Quantification Method dialog box, do the following:
- From Factory Defaults dropdown list, select TMT 10plex.
- Select Create.
- In the Quantification Method Editor dialog box, enter the correction factors.
- Select OK.
- For Save as New Method, enter TMT 10plex phosphopeptide.
- Select Save.