The study holds information necessary to run data analysis.
Procedure
- Select the Start Page tab in Proteome Discoverer.
- On the Start Page, select New Study/Analysis.
- The New Study and Analysis dialog box opens.
- In the New Study and Analysis dialog box, do the following:
- In the Study Name text field, enter a name, for example, TMT Phosphopeptide Example.
- (Optional) For Study Root Directory, change the folder for storing the study.
- Keep the study on a local drive.
- Select the Processing workflow:
- ProcessingWF_Tribrid/PWF_Tribrid_TMT_Quan_SPS_MS3_SequestHT_Percolator.pdProcessingWF.
- Select the Consensus workflow:
- ConsensusWF\CWF_Comprehensive_Enhanced Annotation_Reporter_Quan_MS3.pdConsensusWF.
- Select OK.
- The page for your study opens.
- Define the study factors by doing the following:
- In the Study Definition page, in the Study Factors section, select Add, and select Categorical Factor.
- Select Stimulation as the title of the factor.
- Add the following items to the new study factor: Control, IGF-1, Insulin, and Pool.
- Select Apply.
- The updated study factors appear.
- Add the quantification method to the study.
- In the Quantification Methods section, select Add.
- Select TMT 10plex phosphopeptide.
- The method now appears in the Quantification Methods section.
- Add the raw file to the study:
- Select the Input Files tab.
- Select Add Files.
- In File Explorer, select 20160307_A549_Phospho_1ug_CID_NL_trigger_top1_Lumos_01.raw, and then select OK.
- In the Quan Method column for the raw file, select TMT 10plex phosphopeptide.
- Select OK.
- Drag the file from Input Files to the Files for Analysis area in the Analysis pane.
- Associate the study factors with the quan channels, by doing the following:
- Select the Samples tab.
- In the Stimulation column, make the following changes:
- Change the 126, 127N, and 127C quan channels to Control.
- Change the 128N, 128C, and 129N quan channels to Insulin.
- Change the 129C, 130N, and 130C quan channels to IGF-1.
- Change the 131 quan channel to Pool.
TIP
You can change the value:
- One row at a time by using the dropdown menu.
- Multiple rows at a time by holding down SHIFT or CTRL, right-clicking, and selecting the Set Stimulation option in the menu.