Download the FASTA file using Proteome Discoverer - Proteome Discoverer Software - Download the FASTA file using Proteome Discoverer - Proteome Discoverer Software - Proteome Discoverer Software
Proteome Discoverer 3.1 Familiarization Guide
- Product Tree
- Ardia Platform > Software > Proteome Discoverer Software
- Desktop Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 3.1
- On the Windows desktop, double-click the Proteome Discoverer application.
- On the Start Page, select Administration > Maintain FASTA files.
- In the FASTA Files View, if you have not downloaded FASTA files to the application before, select Check For Updates.
- The Job Queue opens and runs a process that downloads the annotation database from Annotation Server. Wait for this step to complete before proceeding.
- To return to the FASTA Files view, select FASTA Files under Content Management.
- Select Download.
- In the Download from Annotation Server dialog box, do the following.
- For Taxonomy ID, enter 9606.
- Ensure that the Include all subtaxonomies checkbox is clear.
- For Database, select the sp_incl_isoforms database at the top of the list.
- Select Import.
- The Run Queue runs a new process that downloads the human protein database. Depending on your network speed, this process might take a few minutes to complete.
- In the Download from Annotation Server dialog box, do the following.
- For Taxonomy ID, enter 83333.
- Ensure that the Include all subtaxonomies checkbox is clear.
- For Database, select the sp_canonical database at the top of the list.
- Select Import.
- The Run Queue runs a new process that downloads the E. coli protein database. Depending on your network speed, this process might take a few minutes to complete.
- Continue with Create the study.