This section describes using PCA plots and filters.
NOTE
Periodically, the FASTA files provided by the Annotation Server or UniProt are updated. The minor changes in an updated file cause minor changes in the results. The changes are not important in this context. Your results will be nearly identical.
Procedure
- You can access the results by using one of the following methods:
- Select the Analysis Results tab in the study, and do one of the following:
- Double-click the row with the result file.
- Highlight the row and select Open Result to open the result file in a separate tab. - Open the job queue by selecting Administration > Show Job Queue, and do one of the following:
- Double-click the row in the Job Queue for the Consensus workflow that just finished.
- Highlight the row and select Open Results to open the results in a separate tab. - In Windows Explorer, find the study folder, and double-click the result file.