The study holds the study factors, raw data files, quantification methods, and links to all search results for analyses performed on those data files within the study.
Procedure
- Select the Start Page tab in the Proteome Discoverer application.
- Under Start, select New Study/Analysis.
- The New Study and Analysis dialog box opens.
- In the Study Name text box, enter a study name (for example, Triple Knockout Example).
- (Optional) For Study Root Directory, change the folder for storing the study.
- Select the following Processing workflow:
- ProcessingWF_Tribrid\ PWF_Tribrid_TMT_Quan_SPS_MS3_SequestHT_Percolator.pdProcessingWF
- Select the following Consensus workflow:
- ConsensusWF\CWF_Comprehensive_Enhanced Annotation_Reporter_Quan_MS3.pdConsensusWF
- Select OK.
- In the Quantification Methods area on the Study Definition page, select the Add dropdown menu and select TMT 11plex SG253828.
- This is the method you created in Configure the quantification method.
- Define the study factors, and associate them with the quantification channels.
- In the Study Factors area on the Study Definition pane, select the Add dropdown menu and select Categorical Factor.
- For the title of the factor, enter Yeast Strain, and select Apply.
- In the New Factor dialog box, select Edit.
- Add the following items to the new study factor: Met6, His4, Ura2, and Parental.
- Select the Input Files tab, and then select Add Files.
- In the Quan Method column next to the file name, select TMT 11plex SG253828.
- Select and drag the file from the Input Files tab to the Files for Analysis section of the Analysis.
- Select the Samples tab.
- In the Yeast Strain column, make the following changes in the quantification:
- Change the 126, 127N, and 127C quan channels to Met6.
- Change the 128N, 128C, and 129N quan channels to His4.
- Change the 129C, 130N, and 130C quan channels to Ura2.
- Change the 131N and 131C columns to Parental.
TIP
You can enter these changes one row at a time using the dropdown menu, or by selecting multiple rows. Right-click in the dropdown menu and select Set Yeast Strain to.