This familiarization tutorial provides step-by-step instructions for using the Proteome Discoverer application's reporter ion quantification capabilities to analyze three different individual yeast gene knockouts together with controls in a single multiplex LCMS experiment.
This familiarization exercise uses TMT 11plex quantitation. The sample used in this exercise was acquired using Thermo Scientific Orbitrap Fusion™ Lumos™ mass spectrometer.
For general information about TMT-based quantitation, go to https://www.analyteguru.com/.
For information about purchasing the Pierce™ triple knockout yeast standard, go to https://www.thermofisher.com/order/catalog/product/A40938.
Quan method: | TMT 11plex |
|---|---|
Quan channels: | 126, 127N, 127C =ME---T6 128N, 128C, 129N = HIS4 129C, 130N, 130C = URA2 131N, 131C = parental |
Organism: | Yeast |
Instrument: | Orbitrap Fusion Lumos |
Acquisition method: | TMT SPS MS3 (CID MS2, HCD MS3) |
RAW file: | TKOTT11_1ms3_1.raw Located in the application media in the folder: Example RAW Files. Copy to a local directory. |
Sample description: | Three yeast culture replicates each of MET6, HIS4, and URA2 single gene knockout strains as well as two culture replicates of the parental strain using TMT11 chemistry for a single LCMS run |
Disk space: | 1GB local drive space, SSD drive preferred |
Processing time: | Approximately 10 minutes |