The following table contains known defects in the software, categorized by software section, with a brief abstract and information related to each defect’s severity and risk. Product management assesses risk, which can differ significantly from the reported severity.
Software section | Severity | Abstract | Risk |
|---|---|---|---|
Fragment Match View | Medium | For larger proteoforms the fragment match view may load slowly. | Medium |
Workflow management | Medium | FDR cannot be calculated for workflows containing both Med/High and Hi/Hi data types. Split into two workflows. | Low |
ProSightPD Proteoform and Protein Filter | Low | The FDR calculation crashes when using the Medium or High options for proteoform filtering. For this release, the filter was hard-coded to Low. As a workaround, the results in the Proteoform table can be filtered using the Proteoform Characterization Confidence. This will be fixed in the next release. | Low |