If you have a license, but do not have ProSightPD nodes installed, complete the following procedure.
Procedure
- Go to Life Sciences Mass Spectrometry Software Download and Licensing Portal: https://thermo.flexnetoperations.com.
- Log in to your account.
- In the navigation pane, select the Product Search link.
- Enter ProSightPD 4.4.
- In the Product Search Results, select the entry for ProSightPD 4.4.
- Under File Name (right-side column), select the file name link.
After you download and install the software, activate the license with the License Management interface.
If you have issues with licensing, send an email to ThermoMSLicensing@thermofisher.com. Include a short description of the problem and the ProSightPD license key.
To activate the license, you must have an activation code and product ID number from Thermo Fisher Scientific. Before you transfer a license to another computer, deactivate the license. Refer to the Proteome Discoverer User Guide for more information.