Procedure
  1. Select Modify Available PTMs, .
  2. The Global Feature Set Editor opens.
  3. If the modification is not available in Included Features, search for it under Available Features.
  4. Apply a modification on a group or an instance level:
  5. To make the modification group available, in the Available Features list, select the up arrow for the row to move the complete modification group (that is, all the individual instances) to the Included Features list.
  6. To make an instance available, expand the modification group. Select the up arrow on the instance row to move the instance to the Included Features list.
    You can add additional instances one at a time.
  7. (Optional) To globally apply the modification: in the Included Features list, select the modification group or instance, and select the plus icon, .
  8. Select Save.
  9. The new modifications are available.