You can delete a database from the ProSightPD Database Manager if needed.
Prerequisites
- You have the ProSightPD Database Manager open.
Procedure
- In the Manage column, double-click
. - The Delete ProSight Database dialog box opens.
- Select Yes to delete the selected database.
- The database is removed from the list in the Database Manager.