TIP
You can overwrite the existing workflow with the new workflow, that is, you do not need to delete the existing workflow first.
Procedure
  1. Do one of the following:
  2. To use a workflow supplied in the application, select Open Common.
  3. To use a workflow saved on your computer, select Open.
  4. Locate the workflow, and select Open.
  5. (Optional) Repeat to add or change another workflow.
  6. Add files, study factors, and adjust the node parameters to suit your analysis.