Use or edit an analysis template - ProSightPD - Proteome Discoverer Software - ProSightPD 4.4 User Guide - Working with studies - Use or edit an analysis template - Configure a workflow - ProSightPD - ProSightPD - Proteome Discoverer Software - Proteome Discoverer Software
ProSightPD 4.4 User Guide
- Product Tree
- Desktop Software > Proteome Discoverer Software > ProSightPD
- Ardia Platform > Software > Proteome Discoverer Software > ProSightPD
- Desktop Software > Proteome Discoverer Software
- Ardia Platform > Software > Proteome Discoverer Software
- Document Type
- User Guide
- Software version
- 4.4
- Create a study in the dialog box without selecting either a processing or a consensus workflow.
- For instructions, see Create a study.
- On the Study Definition page, select Open Analysis Template
. - In the File Explorer, open the analysis template:
C:\Users\Public\Documents\Thermo\Proteome Discoverer version\Common Templates\ProSightPD version\Analysis Templates - Add files, study factors, and a database and adjust the parameters to suit your analysis.