The ProSightPD application performs the label-free quantification (LFQ) of proteoforms using these nodes:
- ProSightPD Hi Res Feature Detector or the ProSightPD Med Res Feature Detector node in the processing workflow
- Feature Mapper node and ProSightPD Quantifier node in the consensus workflow
The following figure shows the workflows.