A study in the Proteome Discoverer application is a project folder. This folder contains all of the related data files, analyses, results, and meta data for your study.
The ProSightPD application analyzes data by running a processing workflow and a consensus workflow in the study.