The Specialized Traces page provides the total ion chromatogram (TIC) and base peak chromatogram (BPC) for each input file, as shown in the following figure.
The following table lists the columns of the Specialized Traces page.
Column | Description |
---|---|
Tags | Not used with the ProSightPD workflows supplied with the software. |
File ID | Number the application assigns when importing the raw data file. |
Trace/Detector Type | Base Peak Chromatogram (BPC) or Total Ion Chromatogram (TIC). |
Description | MS or MS/MS. |
Scan Filter | The m/z range. |
Spectrum File | The raw data file name. |